Gene Transcripts

Gene Symbol
GPR20
Entrez Gene ID
2843
Gene Name
G protein-coupled receptor 20
Gene Aliases
Chromosome Location
Chr. 8: 141356485 - 141367265 on Build GRCh38
UniGene ID
Hs.152432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005293.2 NP_005284.2 2 152
GenBank mRNA BC117409.1 - 1 156
DR006873.1 - 2 179
JQ308168.1 - 1 63

View Full Product Details