Gene Transcripts

Gene Symbol
HTN1
Entrez Gene ID
3346
Gene Name
histatin 1
Gene Aliases
HIS1
Chromosome Location
Chr. 4: 70050415 - 70058848 on Build GRCh38
UniGene ID
Hs.250959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002159.3 NP_002150.1 5 244
GenBank mRNA BC017835.1 - 5 217
BP335073.2 - 5 221
CB984538.1 - 5 248
CR627329.1 - 3 424
M26664.1 - 4 159

View Full Product Details