Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001329.2 | NP_001001329.1 | 10 | 1315 | |
NM_001289102.1 | NP_001276031.1 | 9 | 1166 | ||
NM_001289103.1 | NP_001276032.1 | 10 | 1319 | ||
NM_001289104.1 | NP_001276033.1 | 10 | 1315 | ||
NM_002743.3 | NP_002734.2 | 10 | 927 | ||
XM_011528130.1 | XP_011526432.1 | 11 | 1081 | ||
XM_011528131.1 | XP_011526433.1 | 11 | 1081 | ||
XM_011528132.1 | XP_011526434.1 | 11 | 1081 | ||
XM_017026977.1 | XP_016882466.1 | 10 | 931 | ||
GenBank mRNA | AF144075.1 | - | 9 | 788 | |
AK130663.1 | - | 10 | 943 | ||
AK225279.1 | - | 10 | 941 | ||
AK225390.1 | - | 10 | 923 | ||
AK290433.1 | - | 9 | 1161 | ||
AK296187.1 | - | 10 | 942 | ||
BC013586.1 | - | 9 | 725 | ||
BF984334.1 | - | 6 | 479 | ||
BT009858.1 | - | 5 | 401 | ||
J03075.1 | - | 10 | 924 | ||
KU178278.1 | - | 5 | 401 | ||
KU178279.1 | - | 5 | 401 |