Gene Transcripts

Gene Symbol
REG1B
Entrez Gene ID
5968
Gene Name
regenerating family member 1 beta
Gene Aliases
PSPS2, REGH, REGI-BETA, REGL
Chromosome Location
Chr. 2: 79085023 - 79088024 on Build GRCh38
UniGene ID
Hs.4158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006507.3 NP_006498.1 6 653
GenBank mRNA BC027895.2 - 5 590
BM310513.1 - 1 152
CA867116.1 - 1 156
D16816.1 - 6 622

View Full Product Details