Gene Transcripts

Gene Symbol
ROM1
Entrez Gene ID
6094
Gene Name
retinal outer segment membrane protein 1
Gene Aliases
ROM, ROSP1, RP7, TSPAN23
Chromosome Location
Chr. 11: 62612741 - 62615120 on Build GRCh38
UniGene ID
Hs.281564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000327.3 NP_000318.1 1 803
GenBank mRNA AK313674.1 - 1 803
BC008100.1 - 1 393 4177529
BC036268.1 - 1 393
L07894.1 - 1 267

View Full Product Details