Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001201359.1 | NP_001188288.1 | 4, 5 | 446 | |
NM_005624.3 | NP_005615.2 | 5 | 449 | ||
XM_011528177.2 | XP_011526479.1 | 6 | 650 | ||
XM_017027119.1 | XP_016882608.1 | 6 | 616 | ||
XM_017027120.1 | XP_016882609.1 | 6 | 545 | ||
XM_017027121.1 | XP_016882610.1 | 6 | 491 | ||
XM_017027122.1 | XP_016882611.1 | 5, 6 | 625 | ||
GenBank mRNA | AB046579.1 | - | 3 | 204 | |
AK292662.1 | - | 4, 5 | 446 | ||
BC125136.1 | - | 3, 4 | 339 | 40119547 | |
BC130561.1 | - | 4 | 384 | 40146592 | |
BC144463.1 | - | 3, 4 | 381 | 9052989 | |
U86358.1 | - | 4 | 338 |