Gene Transcripts
- Gene Symbol
- SPIB
- Entrez Gene ID
- 6689
- Gene Name
- Spi-B transcription factor
- Gene Aliases
- SPI-B
- Chromosome Location
- Chr. 19: 50418938 - 50431053 on Build GRCh38
- UniGene ID
- Hs.437905
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243998.1 |
NP_001230927.1
|
4, 5 |
311 |
|
NM_001243999.1 |
NP_001230928.1
|
5, 6 |
522 |
|
NM_001244000.1 |
NP_001230929.1
|
5, 6 |
491 |
|
NM_003121.4 |
NP_003112.2
|
5, 6 |
526 |
|
GenBank mRNA |
AK225225.1 |
- |
5, 6 |
524 |
|
AK225720.1 |
- |
5, 6 |
526 |
|
AK292644.1 |
- |
5, 6 |
528 |
|
AK300639.1 |
- |
4, 5 |
309 |
|
BC007921.2 |
- |
5, 6 |
505 |
4309499 |
DA957263.1 |
- |
5, 6 |
488 |
|
X66079.1 |
- |
5, 6 |
506 |
|
X96998.1 |
- |
5, 6 |
502 |
|
View Full Product Details