Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003357.4 | NP_003348.1 | 2 | 173 | |
GenBank mRNA | AK312165.1 | - | 2 | 173 | |
BC004481.2 | - | 2 | 157 | 3688615 | |
CR541780.1 | - | 2 | 112 | ||
DB196705.1 | - | 2 | 174 | ||
KU178373.1 | - | 2 | 112 | ||
KU178374.1 | - | 2 | 261 | ||
U01101.1 | - | 2 | 121 | ||
X13197.1 | - | 2 | 119 |