Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284501.1 | NP_001271430.1 | 5 | 726 | |
NM_001284502.1 | NP_001271431.1 | 3 | 337 | ||
NM_012225.3 | NP_036357.1 | 4 | 536 | ||
NR_104318.1 | 3 | 337 | |||
XM_005255027.2 | XP_005255084.1 | 4 | 1003 | ||
XM_005255030.1 | XP_005255087.1 | 3 | 274 | ||
XM_011522338.2 | XP_011520640.1 | 3 | 471 | ||
XM_017022831.1 | XP_016878320.1 | 4 | 670 | ||
XM_017022832.1 | XP_016878321.1 | 3 | 804 | ||
GenBank mRNA | AF118394.1 | - | 4 | 501 | |
AK001023.1 | - | 4 | 512 | ||
AK093593.1 | - | 2 | 1349 | ||
AK300696.1 | - | 3 | 270 | ||
AK315611.1 | - | 4 | 477 | ||
BC002768.2 | - | 4 | 493 | 3633393 | |
BC008005.1 | - | 4 | 518 | 3507768 | |
BI196883.1 | - | 3 | 262 | ||
BU189242.1 | - | 4 | 536 | ||
BX446436.2 | - | 5 | 662 | ||
BX648907.1 | - | 2 | 1304 |