Gene Transcripts

Gene Symbol
DLEU1
Entrez Gene ID
10301
Gene Name
deleted in lymphocytic leukemia 1 (non-protein coding)
Gene Aliases
BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6
Chromosome Location
Chr. 13: 50082169 - 50528643 on Build GRCh38
UniGene ID
Hs.591229
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002605.2
2 597
NR_109973.1
3 650
GenBank mRNA AJ412023.1 - LIT1929
BCMS
2 595
AJ412024.1 - LIT1928
BCMS
3 648
AK095311.1 -
3 570

View Full Product Details