Gene Transcripts
- Gene Symbol
- MYCNOS
- Entrez Gene ID
- 10408
- Gene Name
- MYCN opposite strand
- Gene Aliases
- MYCN-AS1, N-CYM, NCYM, NYCM
- Chromosome Location
- Chr. 2: 15936265 - 15941723 on Build GRCh38
- UniGene ID
- Hs.651453
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026766.1 |
|
1 |
40 |
|
NR_110230.1 |
|
1 |
40 |
|
GenBank mRNA |
S49953.1 |
- |
1 |
40 |
|
- Gene Symbol
- MYCN
- Entrez Gene ID
- 4613
- Gene Name
- v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
- Gene Aliases
- MODED, N-myc, NMYC, ODED, bHLHe37
- Chromosome Location
- Chr. 2: 15940438 - 15947007 on Build GRCh38
- UniGene ID
- Hs.25960
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M55330.1 |
- |
1 |
1240 |
|
View Full Product Details