Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320454.1 | NP_001307383.1 | 3 | 426 | |
NM_006365.2 | NP_006356.1 | 5 | 558 | ||
NR_135263.1 | 4 | 462 | |||
NR_135264.1 | 4 | 462 | |||
NR_135268.1 | 5 | 792 | |||
XM_011509070.1 | XP_011507372.1 | 4 | 871 | ||
XM_011509071.2 | XP_011507373.1 | 4 | 1658 | ||
XM_011509072.1 | XP_011507374.1 | 4 | 481 | ||
XM_011509073.2 | XP_011507375.1 | 3 | 645 | ||
XM_011509074.1 | XP_011507376.1 | 3 | 426 | ||
XM_017000061.1 | XP_016855550.1 | 6 | 709 | ||
XM_017000062.1 | XP_016855551.1 | 8 | 1039 | ||
XM_017000063.1 | XP_016855552.1 | 6 | 663 | ||
XM_017000065.1 | XP_016855554.1 | 7 | 848 | ||
GenBank mRNA | AK124566.1 | - | 1 | 1169 | |
BC050448.1 | - | 4 | 327 | ||
BC059943.1 | - | 4 | 313 | ||
BI668594.1 | - | 4 | 345 | ||
DB534652.2 | - | 2 | 237 | ||
HY139418.1 | - | 3 | 426 | ||
HY158402.1 | - | 4 | 340 | ||
U49857.1 | - | 5 | 402 |