Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015056.2 | NP_055871.1 | 13 | 1664 | |
GenBank mRNA | AK056263.1 | - | 13 | 1627 | |
AK124620.1 | - | 2 | 1213 | ||
BC028386.2 | - | 13 | 1669 | 4821672 | |
BX479134.1 | - | 1 | 301 | ||
D80001.1 | - | 13 | 1562 |