Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
23589
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
CRHSP-24, CRHSP24, CSDC1
Chromosome Location
Chr. 16: 8852942 - 8869012 on Build GRCh38
UniGene ID
Hs.632184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042476.2 NP_001035941.1 2 408
NM_001278260.1 NP_001265189.1 3 216
NM_001278261.1 NP_001265190.1 3 355
NM_001278262.1 NP_001265191.1 3 314
NM_001278263.1 NP_001265192.1 2 90
NM_001278264.1 NP_001265193.1 2 232
NM_001278265.1 NP_001265194.1 2 224
NM_001278266.1 NP_001265195.1 2 200
NM_014316.3 NP_055131.2 2 102
XM_005255228.4 XP_005255285.3 2 266
XM_005255229.4 XP_005255286.1 2 78
XM_011522444.2 XP_011520746.1 2 77
GenBank mRNA AF115345.1 - 1 59
AK093698.1 - 2 96
AK311777.1 - 2 96
BC001333.1 - 2 408
BC003366.1 - 2 99 2900952
BC108283.1 - 2 82 4430943
BM693675.1 - 2 232
BM920489.1 - 2 90
CA396216.1 - 2 159
DA736976.1 - 3 356
DB101255.1 - 3 210
DB446945.1 - 3 277
HY005510.1 - 2 225
HY090855.1 - 2 102

View Full Product Details