Gene Transcripts
- Gene Symbol
- ANAPC13
- Entrez Gene ID
- 25847
- Gene Name
- anaphase promoting complex subunit 13
- Gene Aliases
- APC13, SWM1
- Chromosome Location
- Chr. 3: 134477704 - 134486023 on Build GRCh38
- UniGene ID
- Hs.106909
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242374.1 |
NP_001229303.1
|
2, 3 |
837 |
|
NM_001242375.1 |
NP_001229304.1
|
2, 3 |
422 |
|
NM_015391.3 |
NP_056206.1
|
2, 3 |
203 |
|
GenBank mRNA |
AF086169.1 |
- |
1, 2 |
455 |
|
AK001285.1 |
- |
2, 3 |
834 |
|
AK026550.1 |
- |
2, 3 |
157 |
|
AK290761.1 |
- |
2, 3 |
433 |
|
AL050051.1 |
- |
2, 3 |
422 |
|
AV682398.1 |
- |
2, 3 |
168 |
|
BC005398.1 |
- |
2, 3 |
169 |
3828623 |
BG428689.1 |
- |
2, 3 |
205 |
|
BU852706.1 |
- |
2, 3 |
182 |
|
BX537696.1 |
- |
3, 4 |
623 |
|
CN258858.1 |
- |
2, 3 |
403 |
|
View Full Product Details