Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 21 | 2808 | |
XM_011510931.2 | XP_011509233.1 | 22 | 3122 | ||
XM_011510932.2 | XP_011509234.1 | 22 | 3124 | ||
XM_011510933.2 | XP_011509235.1 | 22 | 3128 | ||
XM_011510934.2 | XP_011509236.1 | 22 | 3128 | ||
XM_017003773.1 | XP_016859262.1 | 22 | 3124 | ||
XM_017003774.1 | XP_016859263.1 | 21 | 2893 | ||
XM_017003775.1 | XP_016859264.1 | 22 | 3126 | ||
XM_017003776.1 | XP_016859265.1 | 21 | 3010 | ||
XM_017003777.1 | XP_016859266.1 | 20 | 2901 | ||
XM_017003778.1 | XP_016859267.1 | 21 | 2888 | ||
XM_017003779.1 | XP_016859268.1 | 22 | 2824 | ||
XM_017003780.1 | XP_016859269.1 | 21 | 2680 | ||
XM_017003781.1 | XP_016859270.1 | 21 | 2670 | ||
XM_017003782.1 | XP_016859271.1 | 17 | 2019 | ||
GenBank mRNA | AF439717.1 | - | 15 | 1711 | |
AK074062.1 | - | 20 | 2416 | ||
AK074075.1 | - | 3 | 318 | ||
BC027939.1 | - | 4 | 646 |