Gene Transcripts
- Gene Symbol
- POMP
- Entrez Gene ID
- 51371
- Gene Name
- proteasome maturation protein
- Gene Aliases
- C13orf12, HSPC014, PNAS-110, UMP1
- Chromosome Location
- Chr. 13: 28659004 - 28678957 on Build GRCh38
- UniGene ID
- Hs.268742
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015932.5 |
NP_057016.1
|
4, 5 |
443 |
|
GenBank mRNA |
AF077200.1 |
- |
4, 5 |
344 |
|
AF125097.1 |
- |
4, 5 |
343 |
|
AF262975.1 |
- |
4, 5 |
287 |
|
AF275807.1 |
- |
4, 5 |
292 |
|
AK312118.1 |
- |
4, 5 |
314 |
|
BC003390.1 |
- |
4, 5 |
306 |
3446986 |
BC014334.1 |
- |
4, 5 |
337 |
4045091 |
EF535527.1 |
- |
4, 5 |
275 |
|
GU045555.1 |
- |
4, 5 |
443 |
|
GU045556.1 |
- |
4, 5 |
442 |
|
View Full Product Details