Gene Transcripts
- Gene Symbol
- HIGD1B
- Entrez Gene ID
- 51751
- Gene Name
- HIG1 hypoxia inducible domain family member 1B
- Gene Aliases
- CLST11240, CLST11240-15
- Chromosome Location
- Chr. 17: 44844281 - 44850480 on Build GRCh38
- UniGene ID
- Hs.151787
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271880.1 |
NP_001258809.1
|
2, 3 |
262 |
|
NM_016438.3 |
NP_057522.1
|
1, 2 |
346 |
|
XM_006721946.2 |
XP_006722009.1
|
2, 3 |
240 |
|
XM_006721947.3 |
XP_006722010.1
|
2, 3 |
650 |
|
XM_006721948.3 |
XP_006722011.1
|
2, 3 |
344 |
|
XM_011524891.2 |
XP_011523193.1
|
2, 3 |
744 |
|
XM_017024742.1 |
XP_016880231.1
|
2, 3 |
690 |
|
XM_017024743.1 |
XP_016880232.1
|
2, 3 |
782 |
|
GenBank mRNA |
AB038021.1 |
- |
1, 2 |
173 |
|
AB038022.1 |
- |
1, 2 |
173 |
|
AW207459.1 |
- |
2, 3 |
295 |
|
BC020667.1 |
- |
1, 2 |
337 |
4724613 |
BG573492.1 |
- |
1, 2 |
338 |
|
BM503772.1 |
- |
1, 2 |
349 |
|
BQ017345.1 |
- |
2, 3 |
296 |
|
View Full Product Details