Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003654.2 | 2 | 851 | ||
NR_004859.1 | 3 | 1409 | |||
GenBank mRNA | AA280726.1 | - | 2 | 113 | |
AF229246.1 | - | 3 | 2669 | ||
AI218957.1 | - | 2 | 113 | ||
AK022844.1 | - | 2 | 851 | ||
AK290489.1 | - | 2 | 861 | ||
AL109690.1 | - | 2 | 1409 | ||
AW341587.1 | - | 3 | 193 | ||
BC011547.1 | - | 2 | 833 | ||
BC012929.1 | - | 2 | 833 | ||
BC063836.1 | - | 2 | 807 | ||
BC093922.1 | - | 2 | 484 | ||
BC093924.1 | - | 2 | 484 | ||
BC143241.1 | - | 2 | 484 | ||
BC143243.1 | - | 2 | 524 | 9051744 |