Gene Transcripts

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
Chromosome Location
Chr. 3: 113828182 - 113947174 on Build GRCh38
UniGene ID
Hs.24583
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172105.1 NP_001165576.1 8 1202
NM_017577.4 NP_060047.3 13 1475
XM_005247546.2 XP_005247603.1 12 1391
XM_005247547.1 XP_005247604.1 13 1437
XM_011512930.1 XP_011511232.1 13 1470
XM_011512931.1 XP_011511233.1 8 1259
XM_017006646.1 XP_016862135.1 12 1389
XM_017006647.1 XP_016862136.1 14 1553
XM_017006648.1 XP_016862137.1 9 1282
XM_017006649.1 XP_016862138.1 7 1033
GenBank mRNA AK093181.1 - 8 994
AK097783.1 - 14 1559
AK292846.1 - 13 1411
AK292964.1 - 8 1117
AK304142.1 - 8 1202
AL133661.1 - 4 1691
AY358725.1 - 8 1155
BC028972.2 - 5 479
BC035040.1 - 13 1481 4829130

View Full Product Details