Gene Transcripts
- Gene Symbol
- KLK15
- Entrez Gene ID
- 55554
- Gene Name
- kallikrein related peptidase 15
- Gene Aliases
- ACO, HSRNASPH
- Chromosome Location
- Chr. 19: 50825289 - 50834337 on Build GRCh38
- UniGene ID
- Hs.567535
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001277081.1 |
NP_001264010.1
|
2, 3 |
219 |
|
NM_001277082.1 |
NP_001264011.1
|
2, 3 |
219 |
|
NM_017509.3 |
NP_059979.2
|
2, 3 |
222 |
|
NR_102274.1 |
|
2, 3 |
219 |
|
XM_006723265.3 |
XP_006723328.1
|
3, 4 |
1023 |
|
XM_011527085.2 |
XP_011525387.1
|
3, 4 |
1019 |
|
XM_011527087.2 |
XP_011525389.1
|
3, 4 |
1027 |
|
XM_011527088.2 |
XP_011525390.1
|
3, 4 |
1033 |
|
XM_011527089.2 |
XP_011525391.1
|
3, 4 |
1030 |
|
XM_017026943.1 |
XP_016882432.1
|
3, 4 |
1033 |
|
GenBank mRNA |
AF303046.1 |
- |
2, 3 |
221 |
|
AY373373.1 |
- |
2, 3 |
191 |
|
AY373374.1 |
- |
2, 3 |
191 |
|
BC069480.1 |
- |
2, 3 |
219 |
7262190 |
BC069507.1 |
- |
2, 3 |
222 |
7262202 |
BC069518.1 |
- |
2, 3 |
219 |
7262214 |
BC144045.1 |
- |
2, 3 |
209 |
|
BC144046.1 |
- |
2, 3 |
209 |
9052566 |
View Full Product Details