Gene Transcripts

Gene Symbol
ANKMY2
Entrez Gene ID
57037
Gene Name
ankyrin repeat and MYND domain containing 2
Gene Aliases
ZMYND20
Chromosome Location
Chr. 7: 16599776 - 16645817 on Build GRCh38
UniGene ID
Hs.157378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020319.2 NP_064715.1 7 1097
XM_011515458.1 XP_011513760.1 8 1224
GenBank mRNA AK001389.1 - 8 1066
AK001740.1 - 7 1035
AK023704.1 - 7 1034
AL050390.1 - 7 1039
BC015453.1 - 1 15
BC035353.1 - 7 1053 5185501

View Full Product Details