Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018070.2 | NP_001018080.1 | 8 | 972 | |
NM_020441.2 | NP_065174.1 | 7 | 950 | ||
GenBank mRNA | AK000860.1 | - | 7 | 933 | |
AK315399.1 | - | 7 | 905 | ||
AL162072.1 | - | 2 | 223 | ||
BC006449.1 | - | 7 | 900 | 3958930 |