Gene Transcripts

Gene Symbol
ZSWIM5
Entrez Gene ID
57643
Gene Name
zinc finger SWIM-type containing 5
Gene Aliases
Chromosome Location
Chr. 1: 45016399 - 45206586 on Build GRCh38
UniGene ID
Hs.656613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020883.1 NP_065934.1 6 1797
XM_011541861.2 XP_011540163.1 6 1805
GenBank mRNA AB040944.1 - 6 1775

View Full Product Details