Gene Transcripts

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
Chromosome Location
Chr. 1: 228208013 - 228381431 on Build GRCh38
UniGene ID
Hs.650039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098623.2 NP_001092093.2 97 22669
NM_001271223.2 NP_001258152.2 108 25540
XM_005273287.4 XP_005273344.1 109 25926
XM_005273291.4 XP_005273348.1 109 25890
XM_005273298.4 XP_005273355.1 108 25617
XM_005273307.4 XP_005273364.1 104 24804
XM_006711822.3 XP_006711885.1 107 25350
XM_006711823.3 XP_006711886.1 107 25344
XM_011544297.2 XP_011542599.1 101 23760
XM_011544299.2 XP_011542601.1 100 23484
XM_017002443.1 XP_016857932.1 109 25971
XM_017002444.1 XP_016857933.1 109 25934
XM_017002445.1 XP_016857934.1 109 25968
XM_017002446.1 XP_016857935.1 109 25968
XM_017002447.1 XP_016857936.1 109 25968
XM_017002448.1 XP_016857937.1 109 25821
XM_017002449.1 XP_016857938.1 108 25708
XM_017002450.1 XP_016857939.1 108 25708
XM_017002451.1 XP_016857940.1 108 25708
XM_017002452.1 XP_016857941.1 108 25708
XM_017002453.1 XP_016857942.1 108 25695
XM_017002454.1 XP_016857943.1 108 25695
XM_017002455.1 XP_016857944.1 108 25695
XM_017002456.1 XP_016857945.1 108 25695
XM_017002457.1 XP_016857946.1 108 25695
XM_017002458.1 XP_016857947.1 107 25444
XM_017002459.1 XP_016857948.1 107 25444
XM_017002460.1 XP_016857949.1 107 25444
XM_017002461.1 XP_016857950.1 107 25431
XM_017002462.1 XP_016857951.1 107 25419
XM_017002463.1 XP_016857952.1 107 25398
XM_017002464.1 XP_016857953.1 107 25347
XM_017002465.1 XP_016857954.1 106 25180
XM_017002466.1 XP_016857955.1 106 25180
XM_017002467.1 XP_016857956.1 106 25180
XM_017002468.1 XP_016857957.1 105 24916
XM_017002469.1 XP_016857958.1 103 24388
XM_017002470.1 XP_016857959.1 98 23069
GenBank mRNA AB046859.2 - 37 6444
AM231061.1 - 23 4571
DQ400343.1 - 23 4571

View Full Product Details