Gene Transcripts

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
Chromosome Location
Chr. 1: 228100727 - 228103462 on Build GRCh38
UniGene ID
Hs.445952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024319.3 NP_077295.1 6 755
GenBank mRNA AY137773.1 - 6 616
BC002843.2 - 6 614 3634226
BC062585.1 - 5 755 5212995

View Full Product Details