Gene Transcripts
- Gene Symbol
- TTTY13
- Entrez Gene ID
- 83868
- Gene Name
- testis-specific transcript, Y-linked 13 (non-protein coding)
- Gene Aliases
- NCRNA00136, TTY13
- Chromosome Location
- Chr. Y: 21583600 - 21594666 on Build GRCh38
- UniGene ID
- Hs.615432
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001537.1 |
|
2, 3 |
244 |
|
View Full Product Details