Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033103.4 | NP_149094.3 | 2 | 156 | |
GenBank mRNA | AF268032.1 | - | 2 | 117 | |
AF423421.1 | - | 2 | 90 | ||
AJ347750.1 | - | 2 | 90 | ||
AK098246.1 | - | 2 | 126 | ||
AK300775.1 | - | 2 | 126 | ||
AK315105.1 | - | 2 | 126 | ||
BC036447.2 | - | 2 | 126 | 4830913 | |
DC348657.1 | - | 2 | 157 |