Gene Transcripts

Gene Symbol
MEX3A
Entrez Gene ID
92312
Gene Name
mex-3 RNA binding family member A
Gene Aliases
MEX-3A, RKHD4
Chromosome Location
Chr. 1: 156072013 - 156081998 on Build GRCh38
UniGene ID
Hs.591496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001093725.1 NP_001087194.1 2 524
GenBank mRNA AK125482.1 - 1 26
AY950677.1 - 2 524

View Full Product Details