Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173512.1 | NP_001166983.1 | 4 | 728 | |
NM_138777.3 | NP_620132.1 | 5 | 884 | ||
NM_199177.2 | NP_954646.1 | 4 | 548 | ||
XM_005252302.3 | XP_005252359.1 | 6 | 952 | ||
XM_011519183.2 | XP_011517485.1 | 6 | 1207 | ||
XM_011519184.2 | XP_011517486.1 | 6 | 717 | ||
XM_011519185.2 | XP_011517487.1 | 6 | 952 | ||
XM_011519186.1 | XP_011517488.1 | 5 | 632 | ||
XM_011519187.2 | XP_011517489.1 | 4 | 672 | ||
GenBank mRNA | AK291603.1 | - | 5 | 574 | |
AK292790.1 | - | 7 | 908 | ||
AK300718.1 | - | 4 | 431 | ||
BC013049.1 | - | 4 | 545 | 3882448 | |
BC053604.1 | - | 4 | 546 | 6008684 |