Gene Transcripts

Gene Symbol
LDHAL6B
Entrez Gene ID
92483
Gene Name
lactate dehydrogenase A like 6B
Gene Aliases
LDH6B, LDHAL6, LDHL
Chromosome Location
Chr. 15: 59206816 - 59208588 on Build GRCh38
UniGene ID
Hs.307052
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033195.2 NP_149972.1 1 1032
GenBank mRNA AK058192.1 - 1 962
AK312777.1 - 1 982
AY009108.1 - 1 1005
AY642121.1 - 1 1025
BC022034.1 - 1 1018 4825988

View Full Product Details