Gene Transcripts

Gene Symbol
C14orf79
Entrez Gene ID
122616
Gene Name
chromosome 14 open reading frame 79
Gene Aliases
Chromosome Location
Chr. 14: 104985801 - 105000310 on Build GRCh38
UniGene ID
Hs.27183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174891.3 NP_777551.2 5 1086
GenBank mRNA AF038188.1 - 5 650
AK057249.1 - 6 1162
BC003539.1 - 4 398
BC011537.1 - 4 916
BC025175.1 - 5 597
BC137495.1 - 5 1029 9021495
BC144671.1 - 5 1015

View Full Product Details