Gene Transcripts

Gene Symbol
LRR1
Entrez Gene ID
122769
Gene Name
leucine rich repeat protein 1
Gene Aliases
4-1BBLRR, LRR-1, PPIL5
Chromosome Location
Chr. 14: 49598697 - 49614672 on Build GRCh38
UniGene ID
Hs.451090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152329.3 NP_689542.2 2 586
NM_203467.1 NP_982292.1 2 586
NR_037792.1 2 586
NR_037793.1 3 694
GenBank mRNA AA742244.1 - 2 452
AK293156.1 - 2 305
AY052405.1 - 2 272
BC030142.2 - 2 522
BC064133.1 - 3 439
BC093697.1 - 2 288
BC107888.1 - 2 335
BC112241.1 - 2 288
BC139921.1 - 2 326 40148567
BI825814.1 - 2 513
BI826606.1 - 2 587
BI829295.1 - 2 523
BQ059807.1 - 2 256
BX248298.1 - 2 329
BX648029.1 - 2 509

View Full Product Details