Gene Transcripts
- Gene Symbol
- C19orf70
- Entrez Gene ID
- 125988
- Gene Name
- chromosome 19 open reading frame 70
- Gene Aliases
- MIC13, P117, QIL1
- Chromosome Location
- Chr. 19: 5678414 - 5680896 on Build GRCh38
- UniGene ID
- Hs.356626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308240.1 |
NP_001295169.1
|
4, 5 |
867 |
|
NM_205767.2 |
NP_991330.1
|
3, 4 |
671 |
|
XM_017026248.1 |
XP_016881737.1
|
3, 4 |
612 |
|
GenBank mRNA |
AA292888.1 |
- |
3, 4 |
438 |
|
AI300708.1 |
- |
1, 2 |
232 |
|
AJ707830.1 |
- |
3, 4 |
417 |
|
BC009557.1 |
- |
3, 4 |
305 |
3899541 |
BC042386.1 |
- |
3, 4 |
280 |
5288954 |
BC052640.1 |
- |
3, 4 |
282 |
|
BI827162.1 |
- |
3, 4 |
670 |
|
View Full Product Details