Gene Transcripts
- Gene Symbol
- RTP1
- Entrez Gene ID
- 132112
- Gene Name
- receptor transporter protein 1
- Gene Aliases
- Z3CXXC1
- Chromosome Location
- Chr. 3: 187197486 - 187201465 on Build GRCh38
- UniGene ID
- Hs.518480
- Species
- Homo sapiens
- Gene Symbol
- LOC101929106
- Entrez Gene ID
- 101929106
- Gene Name
- uncharacterized LOC101929106
- Gene Aliases
- Chromosome Location
- Chr. 3: 187197090 - 187207629 on Build GRCh38
- UniGene ID
- Hs.640145
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110052.1 |
|
3 |
756 |
|
GenBank mRNA |
AK127178.1 |
- |
3 |
756 |
|
View Full Product Details