Gene Transcripts

Gene Symbol
ARL9
Entrez Gene ID
132946
Gene Name
ADP ribosylation factor like GTPase 9
Gene Aliases
Chromosome Location
Chr. 4: 56505209 - 56525174 on Build GRCh38
UniGene ID
Hs.444449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206919.1 NP_996802.1 4 746
XM_005265719.3 XP_005265776.1 4 860
XM_005265720.3 XP_005265777.1 4 928
XM_006714036.3 XP_006714099.1 4 620
GenBank mRNA AY439003.1 - 4 746

View Full Product Details