Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080746.2 | NP_542784.1 | 1 | 341 | |
GenBank mRNA | AB063608.1 | - | 1 | 290 | |
AK130863.1 | - | 1 | 301 | ||
BC014310.1 | - | 1 | 341 | 3935452 | |
BC066312.1 | - | 1 | 297 | 5296757 |