Gene Transcripts

Gene Symbol
APH1B
Entrez Gene ID
83464
Gene Name
aph-1 homolog B, gamma-secretase subunit
Gene Aliases
APH-1B, PRO1328, PSFL, TAAV688
Chromosome Location
Chr. 15: 63277519 - 63309126 on Build GRCh38
UniGene ID
Hs.511703
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145646.1 NP_001139118.1 5 3796
NM_031301.3 NP_112591.2 6 3919
GenBank mRNA AI693802.1 - 1 283
AK057677.1 - 1 2317

View Full Product Details