Gene Transcripts
- Gene Symbol
- NRG4
- Entrez Gene ID
- 145957
- Gene Name
- neuregulin 4
- Gene Aliases
- HRG4
- Chromosome Location
- Chr. 15: 75935190 - 76062997 on Build GRCh38
- UniGene ID
- Hs.732438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138573.3 |
NP_612640.1
|
4, 5 |
422 |
|
XM_017021937.1 |
XP_016877426.1
|
8, 9 |
933 |
|
XM_017021938.1 |
XP_016877427.1
|
7, 8 |
3140 |
|
XM_017021939.1 |
XP_016877428.1
|
7, 8 |
702 |
|
XM_017021940.1 |
XP_016877429.1
|
6, 7 |
3478 |
|
XM_017021941.1 |
XP_016877430.1
|
4, 5 |
441 |
|
XM_017021942.1 |
XP_016877431.1
|
4, 5 |
459 |
|
XM_017021943.1 |
XP_016877432.1
|
4, 5 |
532 |
|
XM_017021944.1 |
XP_016877433.1
|
7, 8 |
3525 |
|
XM_017021945.1 |
XP_016877434.1
|
7, 8 |
3517 |
|
XM_017021946.1 |
XP_016877435.1
|
7, 8 |
3517 |
|
XM_017021947.1 |
XP_016877436.1
|
7, 8 |
3517 |
|
XM_017021948.1 |
XP_016877437.1
|
5, 6 |
3250 |
|
GenBank mRNA |
AK307362.1 |
- |
4, 5 |
400 |
|
AK311507.1 |
- |
5, 6 |
524 |
|
BC017568.1 |
- |
4, 5 |
426 |
4071605 |
BX538100.1 |
- |
5, 6 |
452 |
|
CR749519.1 |
- |
6, 7 |
596 |
|
View Full Product Details