Gene Transcripts

Gene Symbol
FBXO45
Entrez Gene ID
200933
Gene Name
F-box protein 45
Gene Aliases
Fbx45
Chromosome Location
Chr. 3: 196568854 - 196589059 on Build GRCh38
UniGene ID
Hs.169815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105573.1 NP_001099043.1 3 905
GenBank mRNA AK025697.1 - 3 633
BE778263.1 - 3 554

View Full Product Details