Gene Transcripts

Gene Symbol
AMZ2P1
Entrez Gene ID
201283
Gene Name
archaelysin family metallopeptidase 2 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 17: 64966550 - 64975585 on Build GRCh38
UniGene ID
Hs.396447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026903.1 1 336
GenBank mRNA AK056627.1 - 1 319
BX537538.1 - 1 210

View Full Product Details