Gene Transcripts
- Gene Symbol
- SLC29A4
- Entrez Gene ID
- 222962
- Gene Name
- solute carrier family 29 member 4
- Gene Aliases
- ENT4, PMAT
- Chromosome Location
- Chr. 7: 5274311 - 5304073 on Build GRCh38
- UniGene ID
- Hs.4302
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040661.1 |
NP_001035751.1
|
8, 9 |
1184 |
|
NM_001300847.1 |
NP_001287776.1
|
8, 9 |
1139 |
|
NM_153247.2 |
NP_694979.2
|
8, 9 |
1181 |
|
XM_005249658.4 |
XP_005249715.1
|
8, 9 |
1105 |
|
XM_006715667.3 |
XP_006715730.1
|
8, 9 |
1052 |
|
XM_006715668.2 |
XP_006715731.1
|
8, 9 |
1130 |
|
XM_011515200.2 |
XP_011513502.1
|
8, 9 |
1212 |
|
XM_011515201.2 |
XP_011513503.1
|
8, 9 |
1201 |
|
GenBank mRNA |
AK075422.1 |
- |
6, 7 |
758 |
|
AK092242.1 |
- |
8, 9 |
1167 |
|
AY485959.1 |
- |
8, 9 |
1181 |
|
BC025325.1 |
- |
8, 9 |
1040 |
3354432 |
BC047592.1 |
- |
8, 9 |
1145 |
4816109 |
View Full Product Details