Gene Transcripts
- Gene Symbol
- TTTY10
- Entrez Gene ID
- 246119
- Gene Name
- testis-specific transcript, Y-linked 10 (non-protein coding)
- Gene Aliases
- NCRNA00133, TTY10
- Chromosome Location
- Chr. Y: 20465668 - 20519228 on Build GRCh38
- UniGene ID
- Hs.461175
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001542.1 |
|
2 |
1012 |
|
View Full Product Details