Gene Transcripts

Gene Symbol
LYSMD2
Entrez Gene ID
256586
Gene Name
LysM domain containing 2
Gene Aliases
Chromosome Location
Chr. 15: 51723064 - 51751453 on Build GRCh38
UniGene ID
Hs.603629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143917.1 NP_001137389.1 2 463
NM_153374.2 NP_699205.1 2 1054
XM_017022057.1 XP_016877546.1 2 446
GenBank mRNA AK129759.1 - 2 533
AY125955.1 - 2 1043
BC022075.1 - 2 599 4799858
BC033515.1 - 2 1051 5175226
BQ446302.1 - 2 652
CR936638.1 - 1 93

View Full Product Details