Gene Transcripts

Gene Symbol
TMED10P1
Entrez Gene ID
286102
Gene Name
transmembrane p24 trafficking protein 10 pseudogene 1
Gene Aliases
TMED10P, Tmp21-II
Chromosome Location
Chr. 8: 144994865 - 144998897 on Build GRCh38
UniGene ID
Hs.744580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002807.4 1 778

View Full Product Details