Gene Transcripts

Gene Symbol
ZNF252P-AS1
Entrez Gene ID
286103
Gene Name
ZNF252P antisense RNA 1
Gene Aliases
C8orf77
Chromosome Location
Chr. 8: 145002811 - 145006046 on Build GRCh38
UniGene ID
Hs.359978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026974.1 1 720
GenBank mRNA AK092777.1 - 1 545
BC046439.1 - 1 720
BC122544.1 - 1 694

View Full Product Details