Gene Transcripts

Gene Symbol
TRIQK
Entrez Gene ID
286144
Gene Name
triple QxxK/R motif containing
Gene Aliases
C8orf83, PRO0845, UPF0599
Chromosome Location
Chr. 8: 92883530 - 92966150 on Build GRCh38
UniGene ID
Hs.440643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK096398.1 - 1, 2 511
BC104254.1 - 1, 2 359
BC104255.1 - 1, 2 356
BC113007.1 - 1, 2 359
BC113008.1 - 1, 2 359

View Full Product Details