Gene Transcripts
- Gene Symbol
- RBMY1A3P
- Entrez Gene ID
- 286557
- Gene Name
- RNA binding motif protein, Y-linked, family 1, member A3 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Y: 9317061 - 9322874 on Build GRCh38
- UniGene ID
- Hs.567833
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001547.1 |
|
2 |
146 |
|
View Full Product Details