Gene Transcripts

Gene Symbol
KRTAP19-4
Entrez Gene ID
337971
Gene Name
keratin associated protein 19-4
Gene Aliases
KAP19.4
Chromosome Location
Chr. 21: 30496856 - 30497110 on Build GRCh38
UniGene ID
Hs.553693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181610.1 NP_853641.1 1 123
GenBank mRNA AB096945.1 - 1 146
BC117197.1 - 1 167 40125748
BC117199.1 - 1 167 40125750

View Full Product Details