Gene Transcripts

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
Chromosome Location
Chr. 11: 417930 - 442011 on Build GRCh38
UniGene ID
Hs.501622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012302.2 NP_001012302.2 21 2078
XM_011520053.2 XP_011518355.1 21 2174
GenBank mRNA AK026923.1 - 21 2025
AK074088.1 - 14 4695
AK096874.1 - 12 1366
AL832289.1 - 10 2000
BC047946.1 - 2 679
BC128557.1 - 21 2046 40122929
BC144415.1 - 20 2104
U33271.1 - 1 57

View Full Product Details